Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTTTCAGCCTTTAAAAAACAAA[A/C]AAACAAAAAATCTTAAGACATATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605609 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OXR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OXR1 - oxidation resistance 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198532.1 | Intron | NP_001185461.1 | ||||
NM_001198533.1 | Intron | NP_001185462.1 | ||||
NM_001198534.1 | Intron | NP_001185463.1 | ||||
NM_001198535.1 | Intron | NP_001185464.1 | ||||
NM_018002.3 | Intron | NP_060472.2 | ||||
NM_181354.4 | Intron | NP_851999.2 | ||||
XM_006716595.2 | Intron | XP_006716658.1 | ||||
XM_017013589.1 | Intron | XP_016869078.1 | ||||
XM_017013590.1 | Intron | XP_016869079.1 | ||||
XM_017013591.1 | Intron | XP_016869080.1 | ||||
XM_017013592.1 | Intron | XP_016869081.1 | ||||
XM_017013593.1 | Intron | XP_016869082.1 | ||||
XM_017013594.1 | Intron | XP_016869083.1 | ||||
XM_017013595.1 | Intron | XP_016869084.1 | ||||
XM_017013596.1 | Intron | XP_016869085.1 | ||||
XM_017013597.1 | Intron | XP_016869086.1 | ||||
XM_017013598.1 | Intron | XP_016869087.1 | ||||
XM_017013599.1 | Intron | XP_016869088.1 | ||||
XM_017013600.1 | Intron | XP_016869089.1 |