Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609963 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHSY3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CHSY3 - chondroitin sulfate synthase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175856.4 | Intron | NP_787052.3 | ||||
XM_005271982.3 | Intron | XP_005272039.1 | ||||
XM_005271983.3 | Intron | XP_005272040.1 | ||||
XM_011543363.2 | Intron | XP_011541665.1 | ||||
XM_011543364.2 | Intron | XP_011541666.1 | ||||
XM_011543365.2 | Intron | XP_011541667.1 | ||||
XM_017009434.1 | Intron | XP_016864923.1 | ||||
XM_017009435.1 | Intron | XP_016864924.1 |
Set Membership: |
HapMap |