Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142871 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HLA-G PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HLA-G - major histocompatibility complex, class I, G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002127.5 | 675 | Silent Mutation | GGC,GGT | G,G 314 | NP_002118.1 | |
XM_017010816.1 | 675 | Silent Mutation | GGC,GGT | G,G 319 | XP_016866305.1 | |
XM_017010817.1 | 675 | Silent Mutation | GGC,GGT | G,G 222 | XP_016866306.1 | |
XM_017010818.1 | 675 | Silent Mutation | GGC,GGT | G,G 222 | XP_016866307.1 |