Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCGGCGACTGAAGCGCGCGAAAAG[A/C]TGAGGCGGCAACGTCGGGGACGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613644 | ||||||||||||||||||||
Literature Links: |
ATF7IP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATF7IP - activating transcription factor 7 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286514.1 | 110 | UTR 5 | NP_001273443.1 | |||
NM_001286515.1 | 110 | UTR 5 | NP_001273444.1 | |||
NM_018179.4 | 110 | UTR 5 | NP_060649.3 | |||
NM_181352.1 | 110 | Intron | NP_851997.1 | |||
XM_005253424.3 | 110 | Intron | XP_005253481.1 | |||
XM_006719108.3 | 110 | Intron | XP_006719171.1 | |||
XM_006719109.3 | 110 | Intron | XP_006719172.1 | |||
XM_011520754.2 | 110 | Intron | XP_011519056.1 | |||
XM_011520755.2 | 110 | Intron | XP_011519057.1 | |||
XM_011520756.2 | 110 | Intron | XP_011519058.1 | |||
XM_017019638.1 | 110 | Intron | XP_016875127.1 | |||
XM_017019639.1 | 110 | Intron | XP_016875128.1 |