Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGTGAAGAGTGGTGGCCAGAGGC[A/G]GAAGCTAGTGGCCACCCGATGGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
43 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602006 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAPKAPK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAPKAPK2 - mitogen-activated protein kinase-activated protein kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004759.4 | Intron | NP_004750.1 | ||||
NM_032960.3 | Intron | NP_116584.2 | ||||
XM_005273353.3 | Intron | XP_005273410.1 | ||||
XM_017002810.1 | Intron | XP_016858299.1 |
Set Membership: |
HapMap |