Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 605785 | |||||||||||||||||||||||
Literature Links: |
PLEKHH3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PLEKHH3 - pleckstrin homology, MyTH4 and FERM domain containing H3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TUBG2 - tubulin gamma 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320509.1 | 246 | UTR 5 | NP_001307438.1 | |||
NM_016437.2 | 246 | UTR 5 | NP_057521.1 | |||
XM_011524614.2 | 246 | UTR 5 | XP_011522916.1 | |||
XM_017024453.1 | 246 | UTR 5 | XP_016879942.1 | |||
XM_017024454.1 | 246 | UTR 5 | XP_016879943.1 | |||
XM_017024455.1 | 246 | UTR 5 | XP_016879944.1 | |||
XM_017024456.1 | 246 | UTR 5 | XP_016879945.1 | |||
XM_017024457.1 | 246 | UTR 5 | XP_016879946.1 |
Set Membership: |
HapMap |