Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
74 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608729 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGTRAP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AGTRAP - angiotensin II receptor associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040194.1 | Intron | NP_001035284.1 | ||||
NM_001040195.1 | Intron | NP_001035285.1 | ||||
NM_001040196.1 | Intron | NP_001035286.1 | ||||
NM_001040197.1 | Intron | NP_001035287.1 | ||||
NM_020350.4 | Intron | NP_065083.3 | ||||
XM_011541799.2 | Intron | XP_011540101.2 | ||||
XM_011541800.2 | Intron | XP_011540102.1 | ||||
XM_011541802.1 | Intron | XP_011540104.1 |