Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGAGGCTGCTAACTGTGCTCCC[C/T]ACAGCTGGTCTGCAAGTTCATTCAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605002 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CORO2B PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CORO2B - coronin 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190456.1 | Intron | NP_001177385.1 | ||||
NM_001190457.1 | Intron | NP_001177386.1 | ||||
NM_001324014.1 | Intron | NP_001310943.1 | ||||
NM_001324015.1 | Intron | NP_001310944.1 | ||||
NM_006091.4 | Intron | NP_006082.3 |
Set Membership: |
HapMap |