Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602035 MIM: 602427 MIM: 609724 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP4C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PPP4C - protein phosphatase 4 catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303503.1 | 1312 | Intron | NP_001290432.1 | |||
NM_001303504.1 | 1312 | Intron | NP_001290433.1 | |||
NM_001303506.1 | 1312 | Intron | NP_001290435.1 | |||
NM_001303507.1 | 1312 | Intron | NP_001290436.1 | |||
NM_002720.2 | 1312 | Intron | NP_002711.1 | |||
XM_006721061.3 | 1312 | Intron | XP_006721124.1 | |||
XM_017023403.1 | 1312 | Intron | XP_016878892.1 | |||
XM_017023404.1 | 1312 | Intron | XP_016878893.1 |
TBX6 - T-box 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004608.3 | 1312 | Silent Mutation | NP_004599.2 | |||
XM_005255523.2 | 1312 | Silent Mutation | XP_005255580.1 | |||
XM_011545926.2 | 1312 | Silent Mutation | XP_011544228.1 | |||
XM_017023614.1 | 1312 | Silent Mutation | XP_016879103.1 |
YPEL3 - yippee like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |