Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
33 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602516 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RGS4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RGS4 - regulator of G-protein signaling 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102445.2 | Intron | NP_001095915.1 | ||||
NM_001113380.1 | Intron | NP_001106851.1 | ||||
NM_001113381.1 | Intron | NP_001106852.1 | ||||
NM_005613.5 | Intron | NP_005604.1 |
Set Membership: |
HapMap |