Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCACCACGCCTGGCCTTCTGGAA[A/G]CTAATCTCTTATCAGATATATGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602436 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102725068 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC102725068 - uncharacterized LOC102725068 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MICB - MHC class I polypeptide-related sequence B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289160.1 | Intron | NP_001276089.1 | ||||
NM_001289161.1 | Intron | NP_001276090.1 | ||||
NM_005931.4 | Intron | NP_005922.2 |
Set Membership: |
HapMap |