Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604991 MIM: 600129 | ||||||||||||||||||||||||||||||||
Literature Links: |
PART1 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
PART1 - prostate androgen-regulated transcript 1 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDE4D - phosphodiesterase 4D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001104631.1 | Intron | NP_001098101.1 | ||||
NM_001165899.1 | Intron | NP_001159371.1 | ||||
NM_001197218.1 | Intron | NP_001184147.1 | ||||
NM_001197219.1 | Intron | NP_001184148.1 | ||||
NM_001197220.1 | Intron | NP_001184149.1 | ||||
NM_001197221.1 | Intron | NP_001184150.1 | ||||
NM_001197222.1 | Intron | NP_001184151.1 | ||||
NM_001197223.1 | Intron | NP_001184152.1 | ||||
NM_006203.4 | Intron | NP_006194.2 |
Set Membership: |
HapMap |