Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609976 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIF3A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HIF3A - hypoxia inducible factor 3 alpha subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022462.4 | Intron | NP_071907.4 | ||||
NM_152794.3 | Intron | NP_690007.1 | ||||
NM_152795.3 | Intron | NP_690008.2 | ||||
NM_152796.2 | Intron | NP_690009.1 | ||||
XM_005259152.4 | Intron | XP_005259209.1 | ||||
XM_005259153.3 | Intron | XP_005259210.2 | ||||
XM_005259155.4 | Intron | XP_005259212.1 | ||||
XM_005259156.4 | Intron | XP_005259213.1 | ||||
XM_017027132.1 | Intron | XP_016882621.1 | ||||
XM_017027133.1 | Intron | XP_016882622.1 | ||||
XM_017027134.1 | Intron | XP_016882623.1 | ||||
XM_017027135.1 | Intron | XP_016882624.1 | ||||
XM_017027136.1 | Intron | XP_016882625.1 | ||||
XM_017027137.1 | Intron | XP_016882626.1 | ||||
XM_017027138.1 | Intron | XP_016882627.1 | ||||
XM_017027139.1 | Intron | XP_016882628.1 | ||||
XM_017027140.1 | Intron | XP_016882629.1 | ||||
XM_017027141.1 | Intron | XP_016882630.1 | ||||
XM_017027142.1 | Intron | XP_016882631.1 |
Set Membership: |
HapMap |