Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTAGGTAATTTGAATCTTCCTGA[C/T]GAGTCCAGTGATAGTTGATCACTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 190181 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TGFBR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TGFBR1 - transforming growth factor beta receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130916.2 | Intron | NP_001124388.1 | ||||
NM_001306210.1 | Intron | NP_001293139.1 | ||||
NM_004612.3 | Intron | NP_004603.1 | ||||
XM_011518948.2 | Intron | XP_011517250.1 | ||||
XM_011518949.2 | Intron | XP_011517251.1 | ||||
XM_011518950.2 | Intron | XP_011517252.1 | ||||
XM_017015063.1 | Intron | XP_016870552.1 |
Set Membership: |
HapMap |