Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602713 MIM: 610700 MIM: 610081 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADAM9 - ADAM metallopeptidase domain 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003816.2 | 334 | Intron | NP_003807.1 | |||
XM_011544682.2 | 334 | UTR 5 | XP_011542984.1 | |||
XM_017013942.1 | 334 | Intron | XP_016869431.1 |
HTRA4 - HtrA serine peptidase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TM2D2 - TM2 domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024380.1 | 334 | Intron | NP_001019551.1 | |||
NM_001024381.1 | 334 | Intron | NP_001019552.1 | |||
NM_031940.3 | 334 | Intron | NP_114146.3 | |||
NM_078473.2 | 334 | Intron | NP_510882.1 | |||
XM_005273657.3 | 334 | Intron | XP_005273714.1 | |||
XM_006716408.3 | 334 | Intron | XP_006716471.1 | |||
XM_011544667.2 | 334 | Intron | XP_011542969.1 |
Set Membership: |
HapMap |