Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGACCTACCAACACTAGCCCCTC[C/T]CTGGGGGCTCCCAAGTTCTGAAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164010 MIM: 179780 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHMP1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHMP1A - charged multivesicular body protein 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083314.3 | 2177 | UTR 3 | NP_001076783.1 | |||
NM_002768.4 | 2177 | UTR 3 | NP_002759.2 | |||
XM_011523098.1 | 2177 | Intron | XP_011521400.1 |
DPEP1 - dipeptidase 1 (renal) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |