Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605490 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CATSPERD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CATSPERD - cation channel sperm associated auxiliary subunit delta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152784.3 | Intron | NP_689997.3 | ||||
XM_011527885.2 | Intron | XP_011526187.1 | ||||
XM_011527886.2 | Intron | XP_011526188.1 | ||||
XM_011527887.2 | Intron | XP_011526189.1 | ||||
XM_011527888.2 | Intron | XP_011526190.1 | ||||
XM_011527889.2 | Intron | XP_011526191.1 | ||||
XM_011527890.2 | Intron | XP_011526192.1 | ||||
XM_011527891.1 | Intron | XP_011526193.1 | ||||
XM_011527892.1 | Intron | XP_011526194.1 | ||||
XM_017026564.1 | Intron | XP_016882053.1 | ||||
XM_017026565.1 | Intron | XP_016882054.1 | ||||
XM_017026566.1 | Intron | XP_016882055.1 |
LONP1 - lon peptidase 1, mitochondrial | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |