Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194631 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF124 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF124 - zinc finger protein 124 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243740.2 | 635 | UTR 3 | NP_001230669.1 | |||
NM_001297567.1 | 635 | Intron | NP_001284496.1 | |||
NM_001297568.1 | 635 | Intron | NP_001284497.1 | |||
NM_001297569.1 | 635 | Intron | NP_001284498.1 | |||
NM_003431.3 | 635 | Intron | NP_003422.2 | |||
XM_005273257.4 | 635 | Intron | XP_005273314.1 | |||
XM_006711813.3 | 635 | Intron | XP_006711876.1 | |||
XM_011544272.2 | 635 | Intron | XP_011542574.1 |