Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
ALG1L PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ALG1L - ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015050.2 | Intron | NP_001015050.2 | ||||
NM_001195223.1 | Intron | NP_001182152.1 | ||||
XM_017005844.1 | Intron | XP_016861333.1 | ||||
XM_017005845.1 | Intron | XP_016861334.1 | ||||
XM_017005846.1 | Intron | XP_016861335.1 | ||||
XM_017005847.1 | Intron | XP_016861336.1 |
ROPN1B - rhophilin associated tail protein 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012337.2 | Intron | NP_001012337.1 | ||||
NM_001308313.1 | Intron | NP_001295242.1 | ||||
XM_005247138.3 | Intron | XP_005247195.1 | ||||
XM_006713513.3 | Intron | XP_006713576.1 |