Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603241 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LSAMP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LSAMP - limbic system-associated membrane protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318915.1 | 8128 | UTR 3 | NP_001305844.1 | |||
NM_002338.4 | 8128 | UTR 3 | NP_002329.2 | |||
XM_011512840.2 | 8128 | Intron | XP_011511142.1 | |||
XM_017006383.1 | 8128 | Intron | XP_016861872.1 | |||
XM_017006384.1 | 8128 | Intron | XP_016861873.1 |
Set Membership: |
HapMap |