Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604177 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPL8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPL8 - ribosomal protein L8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF517 - zinc finger protein 517 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317936.1 | Intron | NP_001304865.1 | ||||
NM_213605.2 | Intron | NP_998770.2 | ||||
XM_011517014.2 | Intron | XP_011515316.1 | ||||
XM_011517015.2 | Intron | XP_011515317.1 | ||||
XM_011517017.2 | Intron | XP_011515319.1 | ||||
XM_017013384.1 | Intron | XP_016868873.1 |