Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610210 MIM: 611885 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAF1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MAF1 - MAF1 homolog, negative regulator of RNA polymerase III | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SHARPIN - SHANK associated RH domain interactor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR97 - WD repeat domain 97 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316309.1 | Intron | NP_001303238.1 | ||||
XM_017013385.1 | Intron | XP_016868874.1 | ||||
XM_017013386.1 | Intron | XP_016868875.1 | ||||
XM_017013387.1 | Intron | XP_016868876.1 | ||||
XM_017013388.1 | Intron | XP_016868877.1 | ||||
XM_017013389.1 | Intron | XP_016868878.1 | ||||
XM_017013390.1 | Intron | XP_016868879.1 | ||||
XM_017013391.1 | Intron | XP_016868880.1 | ||||
XM_017013392.1 | Intron | XP_016868881.1 | ||||
XM_017013393.1 | Intron | XP_016868882.1 | ||||
XM_017013394.1 | Intron | XP_016868883.1 |