Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCGGCAGGGGGTCGTGCCCACG[G/T]AGCGGGTGGGAACCGAGGCCGCGCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604346 MIM: 612057 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAN1B1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MAN1B1 - mannosidase alpha class 1B member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAN1B1-AS1 - MAN1B1 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SAPCD2 - suppressor APC domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UAP1L1 - UDP-N-acetylglucosamine pyrophosphorylase 1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207309.2 | Intron | NP_997192.2 | ||||
XM_006717317.3 | Intron | XP_006717380.1 | ||||
XM_011519182.2 | Intron | XP_011517484.1 |