Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTGATTTGTTGCTATGAAAGTTA[C/T]TGTATAAGATTTGTGAGGAAAAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
38 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601643 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929541 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101929541 - uncharacterized LOC101929541 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP2R5A - protein phosphatase 2 regulatory subunit B'alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199756.1 | Intron | NP_001186685.1 | ||||
NM_006243.3 | Intron | NP_006234.1 |