Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCACATATGTGTTTGTGTGAGGGG[A/T]GTGTGTGTGTGTGAGAGAGAGAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607770 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLEKHA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLEKHA5 - pleckstrin homology domain containing A5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143821.2 | Intron | NP_001137293.2 | ||||
NM_001190860.2 | Intron | NP_001177789.2 | ||||
NM_001256470.1 | Intron | NP_001243399.1 | ||||
NM_001256787.1 | Intron | NP_001243716.1 | ||||
NM_019012.5 | Intron | NP_061885.2 | ||||
XM_005253400.1 | Intron | XP_005253457.1 | ||||
XM_006719091.1 | Intron | XP_006719154.1 | ||||
XM_006719093.1 | Intron | XP_006719156.1 | ||||
XM_006719096.1 | Intron | XP_006719159.1 | ||||
XM_011520714.1 | Intron | XP_011519016.1 | ||||
XM_011520715.1 | Intron | XP_011519017.1 | ||||
XM_011520716.1 | Intron | XP_011519018.1 | ||||
XM_011520717.1 | Intron | XP_011519019.1 | ||||
XM_017019495.1 | Intron | XP_016874984.1 | ||||
XM_017019496.1 | Intron | XP_016874985.1 | ||||
XM_017019497.1 | Intron | XP_016874986.1 | ||||
XM_017019498.1 | Intron | XP_016874987.1 | ||||
XM_017019499.1 | Intron | XP_016874988.1 | ||||
XM_017019500.1 | Intron | XP_016874989.1 | ||||
XM_017019501.1 | Intron | XP_016874990.1 | ||||
XM_017019502.1 | Intron | XP_016874991.1 | ||||
XM_017019503.1 | Intron | XP_016874992.1 |