Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C19orf33 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C19orf33 - chromosome 19 open reading frame 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317801.1 | 447 | UTR 3 | NP_001304730.1 | |||
NM_033520.2 | 447 | Missense Mutation | AAG,ACG | K,T 96 | NP_277055.1 |
YIF1B - Yip1 interacting factor homolog B, membrane trafficking protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039671.2 | 447 | UTR 3 | NP_001034760.1 | |||
NM_001039672.2 | 447 | UTR 3 | NP_001034761.1 | |||
NM_001039673.2 | 447 | UTR 3 | NP_001034762.1 | |||
NM_001145461.1 | 447 | UTR 3 | NP_001138933.1 | |||
NM_001145462.1 | 447 | UTR 3 | NP_001138934.1 | |||
NM_001145463.1 | 447 | UTR 3 | NP_001138935.1 | |||
XM_005259385.3 | 447 | Intron | XP_005259442.1 | |||
XM_017027449.1 | 447 | Intron | XP_016882938.1 | |||
XM_017027450.1 | 447 | Intron | XP_016882939.1 | |||
XM_017027451.1 | 447 | Intron | XP_016882940.1 |