Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGAATGGAAGCGCTGTTTCAATG[G/T]GAATTATTGCATTGTTGCTGAGATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615098 | ||||||||||||||||||||
Literature Links: |
TTC28 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TTC28 - tetratricopeptide repeat domain 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145418.1 | 8194 | UTR 3 | NP_001138890.1 | |||
XM_005261405.2 | 8194 | UTR 3 | XP_005261462.1 | |||
XM_006724171.3 | 8194 | UTR 3 | XP_006724234.1 | |||
XM_011530018.2 | 8194 | UTR 3 | XP_011528320.1 | |||
XM_011530019.2 | 8194 | Intron | XP_011528321.1 | |||
XM_011530020.1 | 8194 | Intron | XP_011528322.1 | |||
XM_011530021.2 | 8194 | Intron | XP_011528323.1 | |||
XM_011530022.1 | 8194 | Intron | XP_011528324.1 | |||
XM_017028673.1 | 8194 | UTR 3 | XP_016884162.1 |
TTC28-AS1 - TTC28 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |