Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGTTCTGCTAGGTCTTCTGTATT[C/T]TGAGATTTTTCTTCAAGGAGTTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603652 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRPC6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
TRPC6 - transient receptor potential cation channel subfamily C member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004621.5 | 2639 | Silent Mutation | CAA,CAG | Q,Q 904 | NP_004612.2 | |
XM_011542968.2 | 2639 | Silent Mutation | CAA,CAG | Q,Q 849 | XP_011541270.1 | |
XM_017018221.1 | 2639 | Silent Mutation | CAA,CAG | Q,Q 788 | XP_016873710.1 | |
XM_017018222.1 | 2639 | Silent Mutation | CAA,CAG | Q,Q 818 | XP_016873711.1 |
Set Membership: |
HapMap |