Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615693 MIM: 615694 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COLCA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COLCA1 - colorectal cancer associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302644.1 | 257 | Intron | NP_001289573.1 | |||
NM_001302645.1 | 257 | Intron | NP_001289574.1 | |||
NM_001302646.1 | 257 | Intron | NP_001289575.1 | |||
NM_001302647.1 | 257 | Intron | NP_001289576.1 | |||
NM_001302648.1 | 257 | Intron | NP_001289577.1 | |||
NM_207429.3 | 257 | Intron | NP_997312.1 |
COLCA2 - colorectal cancer associated 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136105.2 | 257 | Intron | NP_001129577.1 | |||
NM_001271457.1 | 257 | Intron | NP_001258386.1 | |||
NM_001271458.1 | 257 | Intron | NP_001258387.1 | |||
XM_017017196.1 | 257 | Intron | XP_016872685.1 | |||
XM_017017197.1 | 257 | Intron | XP_016872686.1 | |||
XM_017017198.1 | 257 | Intron | XP_016872687.1 | |||
XM_017017199.1 | 257 | Intron | XP_016872688.1 | |||
XM_017017200.1 | 257 | UTR 5 | XP_016872689.1 |