Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGAAAGACAATCTCTGAGTGGGT[A/G]GGGTGGTGGAAGAGTTTTGGATACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
66 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605763 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC45A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC45A1 - solute carrier family 45 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080397.2 | Intron | NP_001073866.2 | ||||
XM_011541530.1 | Intron | XP_011539832.1 | ||||
XM_011541531.1 | Intron | XP_011539833.1 |
Set Membership: |
HapMap |