Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF708 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF708 - zinc finger protein 708 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297560.1 | 3359 | Intron | NP_001284489.1 | |||
NM_001297561.1 | 3359 | Intron | NP_001284490.1 | |||
NM_021269.2 | 3359 | Intron | NP_067092.2 | |||
XM_017027201.1 | 3359 | Intron | XP_016882690.1 | |||
XM_017027202.1 | 3359 | UTR 3 | XP_016882691.1 | |||
XM_017027203.1 | 3359 | UTR 3 | XP_016882692.1 | |||
XM_017027204.1 | 3359 | UTR 3 | XP_016882693.1 |
Set Membership: |
HapMap |