Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
43 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603418 MIM: 614760 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKR7A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AKR7A2 - aldo-keto reductase family 7 member A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320979.1 | 144 | Intron | NP_001307908.1 | |||
NM_003689.3 | 144 | Intron | NP_003680.2 |
PQLC2 - PQ loop repeat containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040125.1 | 144 | UTR 5 | NP_001035214.1 | |||
NM_001040126.1 | 144 | Intron | NP_001035215.1 | |||
NM_001287531.1 | 144 | Intron | NP_001274460.1 | |||
NM_017765.2 | 144 | Intron | NP_060235.2 | |||
XM_005245916.2 | 144 | Intron | XP_005245973.1 | |||
XM_006710710.2 | 144 | Intron | XP_006710773.1 | |||
XM_006710711.2 | 144 | Intron | XP_006710774.1 | |||
XM_011541634.2 | 144 | Intron | XP_011539936.1 |
Set Membership: |
HapMap |