Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTGGAAAGGTGCTCACATTTGT[C/T]CTCAATGCTCATTGGCTCACATGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616197 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOL10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NOL10 - nucleolar protein 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261392.1 | Intron | NP_001248321.1 | ||||
NM_001261394.1 | Intron | NP_001248323.1 | ||||
NM_024894.3 | Intron | NP_079170.2 | ||||
XM_011510398.2 | Intron | XP_011508700.1 | ||||
XM_011510399.2 | Intron | XP_011508701.1 | ||||
XM_011510400.2 | Intron | XP_011508702.1 | ||||
XM_011510402.2 | Intron | XP_011508704.1 | ||||
XM_017004990.1 | Intron | XP_016860479.1 | ||||
XM_017004991.1 | Intron | XP_016860480.1 |
Set Membership: |
HapMap |