Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGCATCTAGTCGTTCACCATTAA[A/G]TAGGATGGTAGCTGTGGGGTTTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605255 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ETV7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ETV7 - ETS variant 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207035.1 | Intron | NP_001193964.1 | ||||
NM_001207036.1 | Intron | NP_001193965.1 | ||||
NM_001207037.1 | Intron | NP_001193966.1 | ||||
NM_001207038.1 | Intron | NP_001193967.1 | ||||
NM_001207039.1 | Intron | NP_001193968.1 | ||||
NM_001207040.1 | Intron | NP_001193969.1 | ||||
NM_001207041.1 | Intron | NP_001193970.1 | ||||
NM_016135.3 | Intron | NP_057219.1 | ||||
XM_011514659.1 | Intron | XP_011512961.1 |
Set Membership: |
HapMap |