Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0226L PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KIAA0226L - KIAA0226 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286761.1 | 1988 | UTR 3 | NP_001273690.1 | |||
NM_001286762.1 | 1988 | Missense Mutation | CCG,CTG | P,L 629 | NP_001273691.1 | |
NM_001286763.1 | 1988 | UTR 3 | NP_001273692.1 | |||
NM_001286764.1 | 1988 | UTR 3 | NP_001273693.1 | |||
NM_001286765.1 | 1988 | UTR 3 | NP_001273694.1 | |||
NM_001286766.1 | 1988 | UTR 3 | NP_001273695.1 | |||
NM_025113.3 | 1988 | UTR 3 | NP_079389.2 |