Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300137 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGSF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IGSF1 - immunoglobulin superfamily member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170961.1 | Intron | NP_001164432.1 | ||||
NM_001170962.1 | Intron | NP_001164433.1 | ||||
NM_001170963.1 | Intron | NP_001164434.1 | ||||
NM_001555.4 | Intron | NP_001546.2 | ||||
NM_205833.3 | Intron | NP_991402.1 | ||||
XM_011531330.1 | Intron | XP_011529632.1 | ||||
XM_011531333.1 | Intron | XP_011529635.1 | ||||
XM_011531334.1 | Intron | XP_011529636.1 |
Set Membership: |
HapMap |