Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 167409 | ||||||||||||||||||||
Literature Links: |
PAX2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PAX2 - paired box 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000278.4 | Intron | NP_000269.3 | ||||
NM_001304569.1 | Intron | NP_001291498.1 | ||||
NM_003987.4 | Intron | NP_003978.3 | ||||
NM_003988.4 | Intron | NP_003979.2 | ||||
NM_003989.4 | Intron | NP_003980.3 | ||||
NM_003990.4 | Intron | NP_003981.3 |