Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTATTATTGTTGGTTGTTATTACC[A/G]TATTTTGTTATTATTATTACTAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614361 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACSM5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACSM5 - acyl-CoA synthetase medium-chain family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324371.1 | Intron | NP_001311300.1 | ||||
NM_001324372.1 | Intron | NP_001311301.1 | ||||
NM_001324373.1 | Intron | NP_001311302.1 | ||||
NM_017888.2 | Intron | NP_060358.2 | ||||
XM_006721056.3 | Intron | XP_006721119.1 |
PDILT - protein disulfide isomerase like, testis expressed | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |