Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGCATGTAGTAAGCACTTACAA[C/T]TTTTTCCTACATTTTTCTTTTCTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OGFRL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OGFRL1 - opioid growth factor receptor like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324266.1 | Intron | NP_001311195.1 | ||||
NM_024576.4 | Intron | NP_078852.3 | ||||
XM_011536117.2 | Intron | XP_011534419.1 | ||||
XM_011536118.2 | Intron | XP_011534420.1 | ||||
XM_017011303.1 | Intron | XP_016866792.1 | ||||
XM_017011304.1 | Intron | XP_016866793.1 | ||||
XM_017011305.1 | Intron | XP_016866794.1 | ||||
XM_017011306.1 | Intron | XP_016866795.1 |
Set Membership: |
HapMap |