Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605310 MIM: 164177 MIM: 600912 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCHCR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCHCR1 - coiled-coil alpha-helical rod protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105563.1 | 314 | Intron | NP_001099033.1 | |||
NM_001105564.1 | 314 | Intron | NP_001099034.1 | |||
NM_019052.3 | 314 | Intron | NP_061925.2 | |||
XM_011514702.2 | 314 | Intron | XP_011513004.1 | |||
XM_011514703.1 | 314 | Intron | XP_011513005.1 | |||
XM_011514704.2 | 314 | Intron | XP_011513006.1 | |||
XM_011514705.1 | 314 | Intron | XP_011513007.1 | |||
XM_017010961.1 | 314 | Intron | XP_016866450.1 | |||
XM_017010962.1 | 314 | Intron | XP_016866451.1 | |||
XM_017010963.1 | 314 | Intron | XP_016866452.1 | |||
XM_017010964.1 | 314 | Intron | XP_016866453.1 | |||
XM_017010965.1 | 314 | Intron | XP_016866454.1 | |||
XM_017010966.1 | 314 | Intron | XP_016866455.1 | |||
XM_017010967.1 | 314 | Intron | XP_016866456.1 | |||
XM_017010968.1 | 314 | Intron | XP_016866457.1 | |||
XM_017010969.1 | 314 | Intron | XP_016866458.1 | |||
XM_017010970.1 | 314 | Intron | XP_016866459.1 |
POU5F1 - POU class 5 homeobox 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TCF19 - transcription factor 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077511.1 | 314 | Intron | NP_001070979.1 | |||
NM_001318908.1 | 314 | UTR 5 | NP_001305837.1 | |||
NM_007109.2 | 314 | UTR 5 | NP_009040.2 |
Set Membership: |
HapMap |