Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 189907 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNF1B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HNF1B - HNF1 homeobox B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000458.3 | Intron | NP_000449.1 | ||||
NM_001165923.3 | Intron | NP_001159395.1 | ||||
NM_001304286.1 | Intron | NP_001291215.1 | ||||
XM_011525160.1 | Intron | XP_011523462.1 | ||||
XM_011525161.1 | Intron | XP_011523463.1 | ||||
XM_011525162.2 | Intron | XP_011523464.1 | ||||
XM_011525163.2 | Intron | XP_011523465.1 | ||||
XM_011525164.1 | Intron | XP_011523466.1 |
Set Membership: |
HapMap |