Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610990 | ||||||||||||||||||||
Literature Links: |
NRM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NRM - nurim (nuclear envelope membrane protein) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270707.1 | Intron | NP_001257636.1 | ||||
NM_001270708.1 | Intron | NP_001257637.1 | ||||
NM_001270709.1 | Intron | NP_001257638.1 | ||||
NM_001270710.1 | Intron | NP_001257639.1 | ||||
NM_007243.2 | Intron | NP_009174.1 | ||||
XM_017010224.1 | Intron | XP_016865713.1 | ||||
XM_017010225.1 | Intron | XP_016865714.1 |
PPP1R18 - protein phosphatase 1 regulatory subunit 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |