Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGTACTTATCCTCAAGGAGAGT[C/G]AGATATGTGGTTGGTACTTGGGGGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612524 MIM: 605700 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPP21 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RPP21 - ribonuclease P/MRP subunit p21 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM39 - tripartite motif containing 39 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021253.3 | Intron | NP_067076.2 | ||||
NM_172016.2 | Intron | NP_742013.1 |
TRIM39-RPP21 - TRIM39-RPP21 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199119.1 | Intron | NP_001186048.1 |
Set Membership: |
HapMap |