Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604252 MIM: 614263 | ||||||||||||||||||||
Literature Links: |
BACE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BACE1 - beta-secretase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207048.1 | 5733 | UTR 3 | NP_001193977.1 | |||
NM_001207049.1 | 5733 | UTR 3 | NP_001193978.1 | |||
NM_012104.4 | 5733 | UTR 3 | NP_036236.1 | |||
NM_138971.3 | 5733 | UTR 3 | NP_620427.1 | |||
NM_138972.3 | 5733 | UTR 3 | NP_620428.1 | |||
NM_138973.3 | 5733 | UTR 3 | NP_620429.1 |
BACE1-AS - BACE1 antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF214 - ring finger protein 214 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077239.1 | 5733 | Intron | NP_001070707.1 | |||
NM_001278249.1 | 5733 | Intron | NP_001265178.1 | |||
NM_207343.3 | 5733 | Intron | NP_997226.2 |