Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604062 | ||||||||||||||||||||
Literature Links: |
MED16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MED16 - mediator complex subunit 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
R3HDM4 - R3H domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138774.3 | 865 | Missense Mutation | CAG,CGG | Q,R 266 | NP_620129.2 | |
XM_011528416.2 | 865 | Intron | XP_011526718.1 |
RNU6-9 - RNA, U6 small nuclear 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |