Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607951 MIM: 603557 | ||||||||||||||||||||
Literature Links: |
CEP57 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEP57 - centrosomal protein 57 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTMR2 - myotubularin related protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243571.1 | 2490 | UTR 3 | NP_001230500.1 | |||
NM_016156.5 | 2490 | UTR 3 | NP_057240.3 | |||
NM_201278.2 | 2490 | UTR 3 | NP_958435.1 | |||
NM_201281.2 | 2490 | UTR 3 | NP_958438.1 | |||
XM_005274374.2 | 2490 | UTR 3 | XP_005274431.1 | |||
XM_005274375.2 | 2490 | UTR 3 | XP_005274432.1 | |||
XM_006718934.2 | 2490 | UTR 3 | XP_006718997.1 | |||
XM_006718935.2 | 2490 | UTR 3 | XP_006718998.1 | |||
XM_006718936.3 | 2490 | UTR 3 | XP_006718999.1 | |||
XM_011543058.2 | 2490 | UTR 3 | XP_011541360.1 | |||
XM_011543059.2 | 2490 | UTR 3 | XP_011541361.1 | |||
XM_017018517.1 | 2490 | UTR 3 | XP_016874006.1 | |||
XM_017018518.1 | 2490 | UTR 3 | XP_016874007.1 |