Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608426 MIM: 164760 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MKRN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MKRN2 - makorin ring finger protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271707.1 | 2462 | Intron | NP_001258636.1 | |||
NM_014160.4 | 2462 | Intron | NP_054879.3 |
RAF1 - Raf-1 proto-oncogene, serine/threonine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002880.3 | 2462 | UTR 3 | NP_002871.1 | |||
XM_005265355.2 | 2462 | UTR 3 | XP_005265412.1 | |||
XM_005265357.2 | 2462 | UTR 3 | XP_005265414.1 | |||
XM_005265358.4 | 2462 | UTR 3 | XP_005265415.1 | |||
XM_005265359.4 | 2462 | UTR 3 | XP_005265416.1 | |||
XM_011533974.2 | 2462 | UTR 3 | XP_011532276.1 | |||
XM_011533975.2 | 2462 | UTR 3 | XP_011532277.1 | |||
XM_017006966.1 | 2462 | UTR 3 | XP_016862455.1 |