Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAGCGCTTCCTGCTGGAGAGCGA[A/G]CTGCTGCTGCACAGGCAGACAGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928323 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101928323 - uncharacterized LOC101928323 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB47 - zinc finger and BTB domain containing 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145166.3 | 1712 | Silent Mutation | GAA,GAG | E,E 477 | NP_660149.2 |
Set Membership: |
HapMap |