Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAAAGAACAGCATTATGAATGAG[C/T]AGGATAGAACTGATAAAGTAAAACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100419706 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100419706 - zinc finger protein 84 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF577 - zinc finger protein 577 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135590.1 | Intron | NP_001129062.1 | ||||
NM_032679.2 | Intron | NP_116068.2 | ||||
XM_006723432.3 | Intron | XP_006723495.1 | ||||
XM_006723433.2 | Intron | XP_006723496.1 | ||||
XM_011527408.2 | Intron | XP_011525710.1 | ||||
XM_017027387.1 | Intron | XP_016882876.1 |