Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612074 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MGC27345 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MGC27345 - uncharacterized protein MGC27345 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBM28 - RNA binding motif protein 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166135.1 | 6847 | Intron | NP_001159607.1 | |||
NM_018077.2 | 6847 | Intron | NP_060547.2 | |||
XM_011516370.2 | 6847 | UTR 3 | XP_011514672.1 | |||
XM_017012389.1 | 6847 | UTR 3 | XP_016867878.1 | |||
XM_017012390.1 | 6847 | UTR 3 | XP_016867879.1 |